Dihydroartemisinin ≥98.0%
Supplier: TCI America
Synonyms:
α-Dihydroartemisinin
D3793-1G
D3793-5G
TCD3793-1GEA
141.39
USD
TCD3793-1G
TCD3793-5G
Dihydroartemisinin ≥98.0%
Dihydroartemisinin
CAS Number: 71939-50-9
MDL Number: MFCD00274495
Molecular Formula: C15H24O5
Molecular Weight: 284.35
Purity/Analysis Method:
Formula:
C₁₅H₂₄O₅ Melting Pt: 153 °C |
MDL Number:
MFCD00274495 CAS Number: 71939-50-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...