Acrylamide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

A1132-500G A1132-25G
TCA1132-500GEA 81.4 USD
TCA1132-500G TCA1132-025G
Acrylamide ≥98.0% (by titrimetric analysis)
Acrylamide (monomer)

[for Electrophoresis] CAS Number: 79-06-1 MDL Number: MFCD00008032 Molecular Formula: C3H5NO Molecular Weight: 71.08 Purity/Analysis Method: 98.0% (T) Form: Crystal Boiling point (°C): 125 Melting point (°C): 86 Flash Point (°C): 138

Formula: CH₂=CHCONH₂
Boiling Pt: 125 °C
Melting Pt: 86 °C
Density: 1.122 g/cm³ (20 °C)
Flash Pt: 138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008032
CAS Number: 79-06-1
EINECS: 201-173-7
UN: 2074
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db4df01652abb4533fd302fe8c5f2619412914ec&vid=a9f819c9bc16b0cff0b131bd45d9dcb2d0584ffe&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR