3,5-Di-tert-butylsalicylic acid ≥95.0%

Supplier: TCI America

Synonyms: 3,5-Bis-tert-butylsalicylic acid, 3,5-Di-tert-butyl-2-hydroxybenzoic Acid

D1947-25G D1947-500G
TCD1947-025GEA 65 USD
TCD1947-025G TCD1947-500G
3,5-Di-tert-butylsalicylic acid ≥95.0%
3,5-Di-tert-butylsalicylic acid

CAS Number: 19715-19-6 MDL Number: MFCD00059058 Molecular Formula: C15H22O3 Molecular Weight: 250.34 Purity/Analysis Method: 95.0% (T) Form: Crystal Melting point (°C): 163 Flash Point (°C): 179


Other Product Names / Synonyms: 3,5-Di-tert-butyl-2-hydroxybenzoic Acid

Formula: C₁₅H₂₂O₃
MW: 250.34 g/mol
Melting Pt: 163 °C
Flash Pt: 179 °C
MDL Number: MFCD00059058
CAS Number: 19715-19-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=620eb8535cdd68a40b33296925ae910ab0bbb000&vid=620eb8535cdd68a40b33296925ae910ab0bbb000&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR