1-Aminopyrrole ≥98.0%

Supplier: TCI America

Synonyms: 1H-Pyrrol-1-amine

A1022-1G A1022-5G
TCA1022-001GEA 110.9 USD
TCA1022-001G TCA1022-5G
1-Aminopyrrole ≥98.0%
1-Aminopyrrole

CAS Number: 765-39-9 MDL Number: MFCD00059931 Molecular Formula: C4H6N2 Molecular Weight: 82.11 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Boiling point (°C): 174 Specific Gravity (20/20): 1.06 Storage Temperature: 0-10°C

Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 174 °C
Density: 1.06 (20 °C)
MDL Number: MFCD00059931
CAS Number: 765-39-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=105e6bcf4ce401e839b4d4e763bc3c171a480ffd&vid=105e6bcf4ce401e839b4d4e763bc3c171a480ffd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR