Chloroquine diphosphate salt ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Chloroquine phosphate7-Chloro-4-[4-(diethylamino)-1-methylbutylamino]quinoline DiphosphateN4-(7-Chloro-4-quinolyl)-N1,N1-diethyl-1,4-pentanediamine DiphosphateN4-(7-Chloroquinolin-4-yl)-N1,N1-diethylpentane-1,4-diamine bis(phosphate)

C2301-25G C2301-500G C2301-100G
TCC2301-25GEA 99.39 USD
TCC2301-25G TCC2301-500G TCC2301-100G
Chloroquine diphosphate salt ≥98.0% (by HPLC, titration analysis)
Chloroquine diphosphate salt

CAS Number: 50-63-5
MDL Number: MFCD00069852
Molecular Formula: C18H26ClN3
Molecular Weight: 515.86
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 199


Product Categories: Bioscience, Reagents for Pain & Itch Models, Antiprotozoal Ingredients
Other Product Names / Synonyms: N4-(7-Chloro-4-quinolyl)-N1,N1-diethyl-1,4-pentanediamine Diphosphate, 7-Chloro-4-[4-(diethylamino)-1-methylbutylamino]quinoline Diphosphate

Formula: C₁₈H₂₆ClN₃
MW: 515.86 g/mol
Melting Pt: 199 °C
MDL Number: MFCD00069852
CAS Number: 50-63-5
https://us.vwr.com/stibo/web/std.lang.all/93/05/28939305.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR