ɑ-Caryophyllene ≥93.0%
Supplier: TCI America
Synonyms:
alpha-Humulene
C0957-1ML
TCC0957-001MLEA
131.82
USD
TCC0957-001ML
ɑ-Caryophyllene ≥93.0%
alpha-Caryophyllene
CAS Number: 6753-98-6
MDL Number: MFCD00042689
Molecular Formula: C15H24
Molecular Weight: 204.36
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 123
Flash Point (°C): 90
Specific Gravity (20/20): 0.89
Product Categories: Bioscience, Sesquiterpenes
Other Product Names / Synonyms: alpha-Humulene
Formula:
C₁₅H₂₄ MW: 204.36 g/mol Boiling Pt: 123 °C Density: 0.89 (20 °C) Flash Pt: 90 °C |
MDL Number:
MFCD00042689 CAS Number: 6753-98-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...