1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)-1,3-propanedione ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
1-(4-(tert-Butyl)phenyl)-3-(4-methoxyphenyl)propane-1,3-dione
, Avobenzone
B3382-25G
B3382-100G
TCB3382-25GEA
77.69
USD
TCB3382-25G
TCB3382-100G
1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)-1,3-propanedione ≥98.0% (by titrimetric analysis)
1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)-1,3-propanedione
CAS Number: 70356-09-1
MDL Number: MFCD00210252
Molecular Formula: C20H22O3
Molecular Weight: 310.39
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 82
Lambda max.: 356 nm (EtOH)
Formula:
C₂₀H₂₂O₃ MW: 310.39 g/mol Melting Pt: 82 °C |
MDL Number:
MFCD00210252 CAS Number: 70356-09-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...