(2-Bromophenyl)urea ≥98.0% (by titrimetric analysis)

Supplier: TCI America

B2833-5G
TCB2833-5GEA 34.46 USD
TCB2833-5G
(2-Bromophenyl)urea ≥98.0% (by titrimetric analysis)
(2-Bromophenyl)urea

CAS Number: 13114-90-4
Molecular Formula: C7H7BrN2O
Molecular Weight: 215.05
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 202

Formula: C₇H₇BrN₂O
MW: 215.05 g/mol
Melting Pt: 202 °C
MDL Number: MFCD00041315
CAS Number: 13114-90-4
https://us.vwr.com/stibo/web/std.lang.all/43/32/28934332.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR