Bianthrone ≥95.0% (by HPLC)
Supplier: TCI America
CAS Number: 434-85-5
MDL Number: MFCD00001238
Molecular Formula: C28H16O2
Molecular Weight: 384.43
Purity/Analysis Method:
Other Product Names / Synonyms: Dianthrone
Formula:
C₂₈H₁₆O₂ Melting Pt: 300 °C |
MDL Number:
MFCD00001238 CAS Number: 434-85-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...