Trifluoroacetic acid ≥99.0%

Supplier: TCI America
Danger

Synonyms: TFA

T0431-500G T0431-25G T0431-100G
TCT0431-500GEA 177.89 USD
TCT0431-500G TCT0431-025G TCT0431-100G
Trifluoroacetic acid ≥99.0%
Trifluoroacetic acid

CAS Number: 76-05-1 MDL Number: MFCD00004169 Molecular Formula: C2HF3O2 Molecular Weight: 114.02 Purity/Analysis Method: 99.0% (T) Form: Clear Liquid Color: Colorless Boiling point (°C): 73 Specific Gravity (20/20): 1.49


Product Categories: Synthetic Organic Chemistry, C-H Bond Activation Reaction, Peptide Synthesis Other Product Names / Synonyms: TFA

Formula: C₂HF₃O₂
MW: 114.02 g/mol
Boiling Pt: 71.8
Melting Pt: -15
Density: 1.490 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8 I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=871b900083dedee58b0800cd3d6f6036f49984a7&vid=198caec32f48ad15322b0355180067e439d6458f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR