Stearic acid ≥98.0%

Supplier: TCI America

Synonyms: Octadecanoic acid, Octadecylic acid

S0163-25G S0163-500G
TCS0163-025GEA 26.59 USD
TCS0163-025G TCS0163-500G
Stearic acid ≥98.0%
Stearic acid

CAS Number: 57-11-4 MDL Number: MFCD00002752 Molecular Formula: C18H36O2 Molecular Weight: 284.48 Purity/Analysis Method: 98.0% (T) Form: Crystal Color: White Boiling point (°C): 240 Melting point (°C): 70 Flash Point (°C): 182

Formula: C₁₈H₃₆O₂
MW: 284.48 g/mol
Boiling Pt: 361
Melting Pt: 67
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 182 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002752
CAS Number: 57-11-4
EINECS: 200-313-4
Merck Index: 13,08882
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75ff009b9bba4c6d7c28ab3c541f1dcc4c9739f9&vid=f72448c356a8d236535ad340eb7277dd7cec412b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR