Orange G

Supplier: TCI America

Synonyms: Disodium 7-hydroxy-8-phenylazonaphthalene-1,3-disulphonate Acid Orange 10

O0093-25G
TCO0093-025GEA 51.5 USD
TCO0093-025G
Orange G
Orange G

CAS Number: 1936-15-8
MDL Number: MFCD00012457
Molecular Formula: C16H12N2O7S2
Molecular Weight: 452.36
Form: Crystal
Color: Reddish Yellow

Formula: C₁₆H₁₀N₂Na₂O₇S₂
MW: 452.38 g/mol
Melting Pt: 141 °C
Density: 0.45…0.47 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012457
CAS Number: 1936-15-8
EINECS: 217-705-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR