4'-Fluoro-2'-nitroacetanilide ≥98.0%

Supplier: TCI America

Synonyms: N-(4-Fluoro-2-nitrophenyl)acetamide

F0284-25G
TCF0284-025GEA 91 USD
TCF0284-025G
4'-Fluoro-2'-nitroacetanilide ≥98.0%
4'-Fluoro-2'-nitroacetanilide

CAS Number: 448-39-5
MDL Number: MFCD00031517
Molecular Formula: C8H7FN2O3
Molecular Weight: 198.15
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 72

Formula: C₈H₇FN₂O₃
MW: 179.22 g/mol
Melting Pt: 72 °C
MDL Number: MFCD00031517
CAS Number: 448-39-5
https://us.vwr.com/stibo/web/std.lang.all/88/80/28968880.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR