1-Dodecylpyridinium chloride ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: N-Laurylpyridinium chlorideDodecylpyridinium chloride

D0995-25G D0995-500G D0995-100G
TCD0995-500GEA 293.83 USD
TCD0995-500G TCD0995-025G 76218-966
1-Dodecylpyridinium chloride ≥98.0% (by HPLC)
1-Dodecylpyridinium chloride

CAS Number: 104-74-5
MDL Number: MFCD00011987
Molecular Formula: C17H30ClN
Molecular Weight: 283.88
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Flash Point (°C): 165


Product Categories: Materials Chemistry, Cationic Surfactants
Other Product Names / Synonyms: 1-Laurylpyridinium Chloride

Formula: C₁₇H₃₀ClN
MW: 283.88 g/mol
Flash Pt: 165 °C
MDL Number: MFCD00011987
CAS Number: 104-74-5
EINECS: 203-232-2
https://us.vwr.com/stibo/web/std.lang.all/61/74/28976174.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR