Cyclohexyl isocyanide ≥98.0%
Supplier: TCI America
C1438-1G
C1438-5G
TCC1438-1GEA
48.16
USD
TCC1438-1G
TCC1438-005G
Cyclohexyl isocyanide ≥98.0%
Cyclohexyl isocyanide
CAS Number: 931-53-3
MDL Number: MFCD00003839
Molecular Formula: C7H11N
Molecular Weight: 109.17
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 78
Flash Point (°C): 77
Specific Gravity (20/20): 0.90
Product Categories: Synthetic Organic Chemistry, Isonitriles (Multi-component Reaction)
Other Product Names / Synonyms: Isocyanocyclohexane
Formula:
C₇H₁₁N Boiling Pt: 78 °C Density: 0.90 (20 °C) Flash Pt: 77 °C |
MDL Number:
MFCD00003839 CAS Number: 931-53-3 EINECS: 213-238-7 UN: 2929 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...