Sodium hydrogen carbonate concentrate 0.5 M, 100X eluent

Supplier: Ricca Chemical

Synonyms: Sodium bicarbonate

R2805000-50A
RC2805-50AEA 178.63 USD
RC2805-50A
Sodium hydrogen carbonate concentrate 0.5 M, 100X eluent
Sodium hydrogen carbonate

Sodium carbonate eluent

Formula: NaHCO₃
MW: 84.01 g/mol
Density: 1.0000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR