Bottle Assemblies, 1 Liter, Beckman Coulter
Supplier: Beckman Coulter
B29896 B10320 A98813 B29896 A98813 A98812 A98814
76289-242PK 951.29 USD
76289-242 BKB10320_U BKA98813 BKA98812 CA76289-446 BKA98814 CABK363678
Bottle Assemblies, 1 Liter, Beckman Coulter
Bottles Centrifuge Bottles
Beckman Coulter one-litre bottles build upon the versatility of the Avanti Series of centrifuges to help users gain even more control of workflow.
  • Convenient cap handle
  • The only one-litre bottle
  • Approved for use with Avanti
  • Increased speed
  • Easy to use
The only one-litre bottle tested and approved for use with Avanti instruments, the Beckman Coulter one-litre bottle provides operating temperatures, BioSafety, and chemical resistance.
Patented HarvestLine System liners are disposable liners which provide easy-to-use, low-cost sample containment, and are compatible with commonly used chemicals and solvents.
And thanks to convenient features like a cap handle, the new bottle is even easier to work with, so nothing slows down the discovery process.
Ordering information: The Auto Vent Plug O-ring set is designed for use with Beckman Coulter one-litre bottles.
Delivery information: Bottle Assemblies include two bottles, two caps/closures, two auto Vent polypropylene plugs with O-rings, two additional large plug O-rings, and two additional AutoVent plug O-rings. HarvestLine System liner kit includes 120 liners, filling rack, funnel, and instructional CD.