HIGHPlate* 6000 Microplate Rotor, Thermo Scientific
Supplier: Thermo Scientific
75003606 75003606
97040-244EA 9353.3 USD
CA97040-244 97040-244
HIGHPlate* 6000 Microplate Rotor, Thermo Scientific
Centrifuges Rotors
Thermo Scientific* HIGHPlate 6000 Microplate Rotor spins filter plates.
The rotor accommodates a variety of filtration plates up to 80mm high. 2x8 deepwell plates.
Recommended for: DNA purification protocols.
Certifications: Biocontainment certified by CAMR*, HPA, Porton Down, U.K.
Ordering information: Compatible with Thermo Scientific Nalgene bottles and tubes for complete centrifuge solutions. Compatible for Thermo Scientific Multifuge X1/X3/X3 F Series.