RediSep Rf Gold Silica Flash Chromatography Column, Teledyne ISCO
Supplier: Teledyne
692203344 692203359 692203369 692203348 692203347 692203345 692203349 692203346 692203427 692203428
101663-846PK 130.38 USD
101663-846 101663-858 101663-860 101663-854 101663-852 101663-848 101663-856 101663-850 101663-864 101663-866
RediSep Rf Gold Silica Flash Chromatography Column, Teledyne ISCO
Chromatography Columns
RediSep Rf Gold high performance flash columns deliver superior sample purity through the use of fine spherical silica gel (20 to 40 µm)
RediSep Rf Gold normal phase silica offers the capability to improve resolution and isolate difficult compounds such as isomers and impurities. Alternatively, the improved resolution can allow for faster run times or higher loads to save time and solvents. Reusability: Single use.