Sulfanilic acid 0,8% w/v acetic acid_glacial USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

Synonyms: Aniline-4-sulphonic acid, 4-Aminobenzenesulphonic acid, 4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid

R8104000-500C R8104000-120C
RC8104-16EA 82 USD
RC8104-16 RC8104-4
Sulfanilic acid 0,8% w/v acetic acid_glacial USP Test Solution (TS)
Sulphanilic acid
Formula: 4-(H₂N)C₆H₄SO₃H
MW: 173.19 g/mol
Boiling Pt: 363
Melting Pt: 279
Density: 1.0500 g/cm³
Flash Pt: 39 °C (AcOH)
Storage Temperature: Ambient
MDL Number: MFCD00007886
CAS Number: 121-57-3
UN: 2790
ADR: 8 III
Merck Index: 13,09012
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e032bed891f24ee48419dc3c7237b0293fd3e629&vid=28238ab814bb2b8d6eb5012387d8bf41528469e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, Acetic odor
Suitability for Use Pink in presence of Nitrite


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR