Sodium thiosulfate

Supplier: Ricca Chemical
Warning

Synonyms: di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite

R7975000-500A R7975000-4A R7975000-1A
RC7975-16EA 68 USD
RC7975-16 RC7975-1 RC7975-32
Sodium thiosulfate
Sodium thiosulphate
Formula: Na₂S₂O₃
MW: 158.11 g/mol
Boiling Pt: 100
Melting Pt: 48
Density: 1.2300 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
Merck Index: 12,08844

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 136) 2.000 ± 0.001 N at 20.0 °C

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR