Sodium sulfite 5% w/v aqueous solution

Supplier: Ricca Chemical
Danger

R7580000-500A R7580000-1A
RC7580-16EA 81 USD
RC7580-16 RC7580-32
Sodium sulfite 5% w/v aqueous solution
Sodium sulphite
Formula: Na₂SO₃
MW: 126.04 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0400 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS Number: 7757-83-7
Merck Index: 13,08757

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 136) 5.00 ± 0.05 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR