L(+)-Potassium sodium tartrate 25% w/v aqueous solution

Supplier: Ricca Chemical

Synonyms: (R,R)-(+)-Potassium sodium tartrate, L(+)-Tartaric acid potassium sodium salt, Potassium sodium L(+)-tartrate, L-Potassium sodium tartrate, (R,R)-(+)-Tartaric acid potassium sodium salt, (+)-Potassium sodium L-tartrate

R7550000-500A R7550000-120A
RC7550-16EA 120 USD
RC7550-16 RC7550-4
L(+)-Potassium sodium tartrate 25% w/v aqueous solution
L(+)-Potassium sodium tartrate
Formula: C₄H₄KNaO₆
MW: 210.16
Boiling Pt: ~ 100 °C
Density: 1.1164 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00065391
CAS Number: 304-59-6
Merck Index: 12,07840

Order Now

Specification Test Results

Appearance Clear, colorless, odorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR