Sodium peroxodisulfate 1.50 M

Supplier: Ricca Chemical

Synonyms: Disodium peroxodisulphatePeroxydisulphuric acid disodium saltDisodium peroxodisulfateSodium peroxidisulfateSodium peroxidisulphateSodium persulphateSodium persulfatePeroxydisulfuric acid disodium salt

R7516000-20F
RC7516-5EA 784.44 USD
RC7516-5
Sodium peroxodisulfate 1.50 M
Sodium peroxodisulphate
Formula: Na₂S₂O₈
MW: 238.11 g/mol
MDL Number: MFCD00003501
CAS Number: 7775-27-1
UN: 1505
ADR: 5.1,III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 40h) 1.500 ± 0.005 M at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR