Disodium oxalate 0.0250 M (0.0500 N)
Supplier: Ricca Chemical
Synonyms:
Sodium ethanedioate, Sodium oxalate, Soerensen's buffer substances
R7499500-500A
R7499500-1A
RC7499.5-16EA
61
USD
RC7499.5-16
RC7499.5-32
Disodium oxalate 0.0250 M (0.0500 N)
di-Sodium oxalate
Formula:
Na₂C₂O₄ MW: 134 g/mol Boiling Pt: 100 °C Melting Pt: 0.0 °C Density: 1.0020 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00012465 CAS Number: 62-76-0 Merck Index: 13,08723 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 40) | 0.02500 ± 0.00001 M at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...