Sodium hydroxide 0.3743 N
Supplier: Ricca Chemical
Synonyms:
Sodium hydrate
, Soda lye
, Caustic soda
, Lye
R7380000-20F
RC73805EA
711.44
USD
RC73805
Sodium hydroxide 0.3743 N
Sodium hydroxide
Formula:
NaOH MW: 40 g/mol |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 UN: 1823 ADR: 8,II |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 84) | 0.3743 ± 0.0001 N at 20.0 °C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...