Sodium carbonate 0.0200 N (N/50)

Supplier: Ricca Chemical

Synonyms: Calcined sodaCarbonic acid disodium saltNa-XCarbonic acid sodium saltDisodium carbonateSodaBisodium carbonateCarbonic acid, disodium saltSoda ash

R7175000-500A R7175000-4A R7175000-1A
RC7175-1EA 210.56 USD
RC7175-1 RC7175-16 RC7175-32
Sodium carbonate 0.0200 N (N/50)
Sodium carbonate

1 ml = 1 mg CaCO₃

Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0011 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
https://us.vwr.com/stibo/web/std.lang.all/05/89/16640589.jpg

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 723) 0.02000 ± 0.00005 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR