Sodium acetate 2.00 M (2.00 N)

Supplier: Ricca Chemical

Synonyms: Acetic acid sodium salt

R7130000-500A R7130000-4A R7130000-1A
RC7130-16EA 182 USD
RC7130-16 RC7130-1 RC7130-32
Sodium acetate 2.00 M (2.00 N)
Sodium acetate
Formula: H₃CCOONa
MW: 82.03 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0805 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay 2.00 ± 0.06 M

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR