Simulated intestinal fluid USP Test Solution (TS)
Supplier: Ricca Chemical
|
Danger
|
R7109000-500A
R7109000-1A
RC7109-16EA
127
USD
RC7109-16
RC7109-32
Simulated intestinal fluid USP Test Solution (TS)
Simulated intestinal fluid
Specification Test Results
Appearance | Clear to hazy with stringy fibers, colorless, characteristic odor |
pH at 25 °C (traceable to NIST SRM 185 & 186) | 6.8 ± 0.1 pH at 25.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...