Silver nitrate 0.280 N

Supplier: Ricca Chemical

Synonyms: Silver mononitrateNitric acid silver saltLunar caustic

R7010000-1B
RC701032EA 390.61 USD
RC701032
Silver nitrate 0.280 N
Silver nitrate
Formula: AgNO₃
MW: 169.87 g/mol
MDL Number: MFCD00003414
CAS Number: 7761-88-8
UN: 1493
ADR: 5.1,II
https://us.vwr.com/stibo/web/std.lang.all/06/45/16640645.jpg

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 0.2800 ± 0.0001 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR