Potassium sulfate 1% w/v aqueous solution USP Test Solution (TS)

Supplier: Ricca Chemical

R6540000-120A R6540000-500A
RC6540-4EA 59 USD
RC6540-4 RC6540-16
Potassium sulfate 1% w/v aqueous solution USP Test Solution (TS)
Potassium sulphate
Formula: K₂SO₄
MW: 174.26 g/mol
Boiling Pt: 1698
Melting Pt: 1069
Density: 1.0080 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS Number: 7778-80-5
Merck Index: 13,07759

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Refractive Index Confirmed concentration


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR