Ammonia solution 16,7%, Electronic Grade
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
R0632000-1A
R0632000-20F
R0632000-4A
R0632000-500A
RC632-32EA
111
USD
RC632-32
RC632-5
RC632-1
RCR0632000500A
Ammonia solution 16,7%, Electronic Grade
Ammonia solution
Formula:
NH₃ MW: 35.05 g/mol Boiling Pt: ~ 36 °C Melting Pt: ~ 0 °C Density: 0.9820 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00066650 CAS Number: 1336-21-6 UN: 3266 ADR: 8 III |
Specification Test Results
Appearance | Colorless liquid |
Assay (vs. Sulfuric Acid/Methyl Red) | 2.3 - 2.5 N at 20 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...