Potassium iodide 16,5% (w/v) in aqueous solution stabilised USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

Synonyms: Dipotassium diiodide, Kali iodide

R6307000-4B R6307000-500B R6307000-1B
RC6307-1EA 482 USD
RC6307-1 RC6307-16 RC6307-32
Potassium iodide 16,5% (w/v) in aqueous solution stabilised USP Test Solution (TS)
Potassium iodide
Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330
Melting Pt: 686
Density: 1.1200 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
Merck Index: 13,07727

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 16.5 ± 0.1 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR