Potassium hydroxide 0.500 N (N/2) isopropanol

Supplier: Ricca Chemical

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

R6257500-1A R6257500-4A
RC6257.5-32EA 109 USD
RC6257.5-32 RC6257.5-1
Potassium hydroxide 0.500 N (N/2) isopropanol
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1813
ADR: 8 II
Merck Index: 12,07806

Order Now

Specification Test Results

Appearance Clear, colorless, Isopropanol odor
Assay at 20 °C (traceable to NIST SRM 723) 0.5000 ± 0.0005 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR