Potassium hydroxide 0.400 Normal, Tightrant™ Tested in an ISO 17025 Accredited Facility

Supplier: Ricca Chemical

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

R6225000-4A R6225000-1A
RC6225-1EA 125 USD
RC6225-1 RC6225-32
Potassium hydroxide 0.400 Normal, Tightrant™ Tested in an ISO 17025 Accredited Facility
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1813
ADR: 8 II
Merck Index: 12,07806

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 0.4000 ± 0.0002 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR