Potassium hydroxide 0.0100 N (N/100)
Supplier: Ricca Chemical
Synonyms:
Potash caustic, Potassium hydrate, Caustic potash
R6196000-1A
R6196000-500A
RC6196-32EA
72
USD
RC6196-32
RC6196-16
Potassium hydroxide 0.0100 N (N/100)
Potassium hydroxide
Formula:
KOH MW: 56.11 g/mol Boiling Pt: 1320 Melting Pt: 360 |
MDL Number:
MFCD00003553 CAS Number: 1310-58-3 UN: 1813 ADR: 8 II Merck Index: 12,07806 |
Specification Test Results
Appearance | Clear, colorless, odorless |
Assay at 20 °C (traceable to NIST SRM 723) | 0.01000 ± 0.00005 N at 20.0 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...