Potassium hydrogen diiodate 0.100 N (N/10)

Supplier: Ricca Chemical

Synonyms: Potassium bi-iodate

R5879000-1C
RC587932EA 157.13 USD
RC587932
Potassium hydrogen diiodate 0.100 N (N/10)
Potassium hydrogen diiodate
Formula: HI₂KO₆
MW: 389.91
MDL Number: MFCD00011400
CAS Number: 13455-24-8
UN: 1479
ADR: 5.1,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.1000 ± 0.0001 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR