Potassium hydrogen diiodate 0.00564 N

Supplier: Ricca Chemical

Synonyms: Potassium bi-iodate

R5873000-1C
RC5873-32EA 114.45 USD
RC5873-32
Potassium hydrogen diiodate 0.00564 N
Potassium hydrogen diiodate

1 mL = 0.2 mg Cl2 200 ppm

Formula: HI₂KO₆
MW: 389.91
MDL Number: MFCD00011400
CAS Number: 13455-24-8
UN: 1479
ADR: 5.1,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.005640 ± 0.000002 N at 20.0 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR