Aluminium potassium sulfate dodecahydrate 10%

Supplier: Ricca Chemical

Synonyms: Aluminium Potassium sulphate, 12-Hydrate, Aluminium Potassium sulfate, 12-Hydrate, Aluminum potassium sulfate dodecahydrate, Aluminum potassium sulphate dodecahydrate, Aluminium potassium bis(sulphate) dodecahydrate, Potassium alum, Alum potassium, Aluminium potassium bis(sulfate) dodecahydrate

R0558000-500A R0558000-1A
RC558-16EA 101 USD
RC558-16 RC558-32
Aluminium potassium sulfate dodecahydrate 10%
Aluminium potassium sulphate dodecahydrate

Alum, 10% (w/v)

Formula: AlK(SO₄)₂·12H₂O
MW: 474.39 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: 92.5
Density: 1.0100 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149143
CAS Number: 7784-24-9
Merck Index: 12,00373

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Alum Solution APHA (4500-O E)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR