Alkaline pyrogallol reagent for Orsat gas analysis

Supplier: Ricca Chemical
Danger

R0557000-1B R0557000-500B
RC557-32EA 159 USD
RC557-32 RC557-16
Alkaline pyrogallol reagent for Orsat gas analysis
Alkaline pyrogallol reagent
Boiling Pt: ~ 102 °C
Density: 1.3382 g/cm³
Storage Temperature: Ambient
UN: 1814
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, dark brown, odorless


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR