Nitric acid, dilute R 125 g/L
Supplier: Ricca Chemical
|
Danger
|
R5330000-500C
R5330000-1C
RC5330-16EA
85
USD
RC5330-16
RC5330-32
Nitric acid, dilute R 125 g/L
Nitric acid, dilute R
Formula:
HNO3 Boiling Pt: ~ 100 °C Melting Pt: ~ 0 °C Density: 1.0660 g/cm³ Storage Temperature: Ambient |
UN:
3264 ADR: 8,III |
Specification Test Results
Appearance | Clear, colorless, Nitric odor |
Assay at 20 °C (traceable to NIST SRM 84) | 125.0 ± 0.1 g/L HNO3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...