Ninhydrin solution R Reag. Ph. Eur.

Supplier: Ricca Chemical
Danger

Synonyms: 2,2-Dihydroxyindan-1,3-dionTrioxohydrindene monohydrateNinhydrin monohydrate2,2-Dihydroxy-1,3-indanedioneIndanetrione hydrate2,2-Dihydroxyindane-1,3-dione1,2,3-Indantrione monohydrate

R5309000-500C R5309000-120C
RC5309-16EA 312.76 USD
RC5309-16 RC5309-4
Ninhydrin solution R Reag. Ph. Eur.
Ninhydrin
Formula: C₉H₆O₄
MW: 178.14 g/mol
Boiling Pt: ~ 118 °C
Density: 0.8218 g/cm³
Flash Pt: ~ 36…38 °C
Storage Temperature: Ambient
MDL Number: MFCD00003791
CAS Number: 485-47-2
UN: 1120
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear, light yellow, alcohol odor
Suitability for Use Blue color in presence of amino acids, proteins and peptides

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR