1-Naphtholbenzeine 0.25% (w/v) in glacial acetic acid USP Test Solution (TS)
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
alpha-Naphtholbenzein
, α-Naphtholbenzein
, 4-(α-(4-Hydroxy-1-naphthyl)benzylidene)naphthalen-1(4H)-one
, 4-Hydroxy-α-(4-hydroxynaphthyl)-α-phenylnaphthalene-1-methanol
, α-Naphtholbenzeine
, p-Naphtholbenzeine
, 1-Naphtholbenzein
, Bis(4-hydroxy-1-naphthyl)phenylmethanol
, p-Naphtholbenzein
R5227000-120C
R5227000-500C
RC5227-4EA
91.27
USD
RC5227-4
RC5227-16
1-Naphtholbenzeine 0.25% (w/v) in glacial acetic acid USP Test Solution (TS)
1-Naphtholbenzeine
Formula:
C₂₇H₁₈O₂ MW: 374.44 g/mol Boiling Pt: ~ 118 °C Melting Pt: 16.64 °C Density: 1.0500 g/cm³ Flash Pt: 39 °C Storage Temperature: Ambient |
MDL Number:
MFCD00078492 CAS Number: 145-50-6 UN: 2789 ADR: 8 (3),II |
Specification Test Results
Appearance | Dark red, strong acetic odor |
Suitability for Use | Orange (pH 10.0) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...