Mercury(II) thiocyanate stock solution 4,17 g/L in methanol

Supplier: Ricca Chemical

Synonyms: Mercury dithiocyanate, Mercuric thiocyanate, Mercury (II) rhodanide

R4785000-4C R4785000-500C R4785000-1C
RC4785-1EA 287 USD
RC4785-1 RC4785-16 RC4785-32
Mercury(II) thiocyanate stock solution 4,17 g/L in methanol
Mercury (II) thiocyanate
Formula: C₂HgN₂S₂
MW: 316.76 g/mol
Melting Pt: -38.9
Flash Pt: 120
MDL Number: MFCD00011039
CAS Number: 592-85-8
UN: 1646
ADR: 6.1 II
Merck Index: 13,05917

Order Now

Specification Test Results

Appearance Clear, colorless, Methanol odor
Suitability for Use Sensitivity to Chloride


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR