Mercuric sulfate Denigès' reagent USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

R4779000-500A R4779000-120A
RC4779-16EA 238 USD
RC4779-16 RC4779-4
Mercuric sulfate Denigès' reagent USP Test Solution (TS)
Mercuric sulfate Denigès' reagent
Density: 1.2956 g/cm³
Storage Temperature: Ambient
UN: 2922
ADR: 8 (6.1),II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 0.491 ± 0.010 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR