Magnesium sulfate 12% w/v aqueous solution USP Test Solution (TS)

Supplier: Ricca Chemical

Synonyms: MgSO4, Magnesium sulfate anhydrous, Magnesium sulphate anhydrous

R4548000-500A R4548000-1A
RC4548-16EA 49 USD
RC4548-16 RC4548-32
Magnesium sulfate 12% w/v aqueous solution USP Test Solution (TS)
Magnesium sulphate
Formula: MgSO₄
MW: 120.37 g/mol
Boiling Pt: ~ 100 °C
Melting Pt: 1127
Density: 1.0570 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 915) 12.0 ± 0.1 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR