Magnesium chloride 10 g/L in mixed alcohol solvent for determination of salts in crude oil

Supplier: Ricca Chemical
Danger

R4465000-1C
RC4465-32EA 130.12 USD
RC4465-32
Magnesium chloride 10 g/L in mixed alcohol solvent for determination of salts in crude oil
Magnesium chloride
Formula: MgCl₂
MW: 95.21 g/mol
Boiling Pt: ~ 118 °C
Density: 0.8700 g/cm³
Flash Pt: ~ 29 °C
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear, colorless, Butanol odor

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR