Eriochrome cyanine R USP Test Solution (TS)
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Chromoxane Cyanine R
R2903000-500A
R2903000-120A
RC2903-16EA
124
USD
RC2903-16
RC2903-4
Eriochrome cyanine R USP Test Solution (TS)
Eriochrome cyanine R
Formula:
C₂₃H₁₅Na₃O₉S Boiling Pt: ~ 100 °C Melting Pt: ~ 0 °C Density: 1.0100 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00012411 CAS Number: 3564-18-9 |
Specification Test Results
Appearance | Clear, red, odorless |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...