1,5-Diphenylcarbazone solution 1% (w/v) alcoholic solution USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

Synonyms: sym-Diphenylcarbazone, Phenylazoameisensäure-2-phenylhydrazid, Phenylazoformic acid 2-phenylhydrazid, Phenylazoformic acid 2-phenylhydrazide

R2605000-120C R2605000-500C
RC2605-4EA 70 USD
RC2605-4 RC2605-16
1,5-Diphenylcarbazone solution 1% (w/v) alcoholic solution USP Test Solution (TS)
1,5-Diphenylcarbazone
Formula: C₁₃H₁₂N₄O
MW: 240.26 g/mol
Boiling Pt: 440
Melting Pt: 157
Density: 0.8000 g/cm³
Flash Pt: ~ 15.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003024
CAS Number: 538-62-5
UN: 1987
ADR: 3 II
Merck Index: 12,03380

Order Now

Specification Test Results

Appearance Red, alcohol odor
Suitability for Use Colorless (excess Chloride) - Purple (excess Mercury)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR