Calcium chloride 7,5% (w/v) in aqueous solution USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

R1783000-500A R1783000-1A
RC1783-16EA 93 USD
RC1783-16 RC1783-32
Calcium chloride 7,5% (w/v) in aqueous solution USP Test Solution (TS)
Calcium chloride
Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600
Melting Pt: 772
Density: 1.0456 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
Merck Index: 14,01659

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 915) 7.50 ± 0.01 % (w/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR